2,4,5-Trifluorbenzamidin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 2,4,5-Trifluorbenzoesäureamidin Hydrochlorid, 2,4,5-Trifluorphenylamidin Hydrochlorid

12-0063.500 12-0063.1
CHMP12-0063.500EA 0 EUR
CHMP12-0063.500 CHMP12-0063.1
2,4,5-Trifluorbenzamidin Hydrochlorid
2,4,5-Trifluorbenzamidin Hydrochlorid
Formel: C₇H₆ClF₃N₂
Molecular Weight: 210,59 g/mol
MDL: MFCD04114446
CAS-Nummer: 764648-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR