2,4,5-Triiodimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
2,4,5-Triiod-1H-imidazol
HI-1003-5G
HI-1003-100G
HI-1003-25G
COBBHI-1003-5GEA
79.1
EUR
COBBHI-1003-5G
COBBHI-1003-100G
COBBHI-1003-25G
2,4,5-Triiodimidazol
2,4,5-Triiodoimidazole
Formel:
C₃HI₃N₂ Molecular Weight: 445.77 g/mol |
MDL:
MFCD00068812 CAS-Nummer: 1746-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...