2,4,6-Trifluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2,4,6-Trifluorophenyl)metaboric acid, 2,4,6-Trifluorobenzeneboronic acid, Dihydroxy(2,4,6-trifluorophenyl)borane
BB-2674-5G
BB-2674-1G
COBBBB-2674-5GEA
46.2
EUR
COBBBB-2674-5G
COBBBB-2674-1G
2,4,6-Trifluorphenylboronsäure
2,4,6-Trifluorphenylboronsäure
Formel:
C₆H₄BF₃O₂ Molecular Weight: 175,9 g/mol |
MDL:
MFCD01863169 CAS-Nummer: 182482-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...