2,4-Dibrom-1,5-dimethylimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
2,4-Dibrom-1,5-dimethyl-1H-imidazol
HI-1173-1G
HI-1173-5G
COBBHI-1173-1GEA
225
EUR
COBBHI-1173-1G
COBBHI-1173-5G
2,4-Dibrom-1,5-dimethylimidazol
2,4-Dibromo-1,5-dimethylimidazole
Formel:
C₅H₆Br₂N₂ Molecular Weight: 253,92 g/mol |
MDL:
MFCD06659899 CAS-Nummer: 850429-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...