2,4-Dibrom-1-methylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 2,4-Dibrom-1-methyl-1H-imidazol

HI-1189-5G HI-1189-1G
COBBHI-1189-5GEA 1130 EUR
COBBHI-1189-5G COBBHI-1189-1G
2,4-Dibrom-1-methylimidazol
2,4-Dibromo-1-methylimidazole
Formel: C₄H₄Br₂N₂
Molecular Weight: 239,9 g/mol
MDL: MFCD06659906
CAS-Nummer: 53857-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR