2,4-Dibromimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 2,5-Dibromo-1H-imidazole

HI-1029-1G HI-1029-5G HI-1029-25G
COBBHI-1029-1GEA 276 EUR
COBBHI-1029-1G COBBHI-1029-5G COBBHI-1029-25G
2,4-Dibromimidazol
2,4-Dibromoimidazole
Formel: C₃H₂Br₂N₂
Molecular Weight: 225,87 g/mol
MDL: MFCD02179517
CAS-Nummer: 64591-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR