2,4-Dichlor-5-methylbenzolsulfonylchlorid
Lieferant: Thermo Scientific
Synonyme:
2,4-Dichlor-5-methylbenzolsulfochlorid, 2,4-Dichlor-5-methylbenzolsulfonsäurechlorid, 2,4-Dichlor-5-methylbenzol-1-sulfonylchlorid
SB00644.1
SB00644.50
MAYBSB00644.1EA
0
EUR
MAYBSB00644.1
MAYBSB00644.50
2,4-Dichlor-5-methylbenzolsulfonylchlorid
2,4-Dichlor-5-methylbenzolsulfonylchlorid
Formel:
C₇H₅Cl₃O₂S Molecular Weight: 259,54 g/mol |
MDL:
MFCD00044016 CAS-Nummer: 28286-86-4 EINECS: 248-938-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...