2,4-Dichlor-6-methylbenzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 2,4-Dichlor-6-methylbenzol-1-sulfonylchlorid, 2,4-Dichlor-6-methylbenzolsulfonsäurechlorid, 2,4-Dichlor-6-methylbenzolsulfochlorid

TL00692.1 TL00692.10 TL00692.50
MAYBTL00692.1EA 0 EUR
MAYBTL00692.1 MAYBTL00692.10 MAYBTL00692.50
2,4-Dichlor-6-methylbenzolsulfonylchlorid
2,4-Dichlor-6-methylbenzolsulfonylchlorid
Formel: C₇H₅Cl₃O₂S
Molecular Weight: 259.54
MDL: MFCD00204179
CAS-Nummer: 175278-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR