2,4-Dichlorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2,4-Dichlorbenzolboronsäure, Dihydroxy(2,4-dichlorphenyl)boran, (2,4-Dichlorphenyl)metaborsäure
BB-2267-5G
BB-2267-25G
BB-2267-1G
BB-2267-100G
COBBBB-2267-5GEA
35.9
EUR
COBBBB-2267-5G
COBBBB-2267-25G
COBBBB-2267-1G
COBBBB-2267-100G
2,4-Dichlorphenylboronsäure
2,4-Dichlorphenylboronsäure
Formel:
C₆H₅BCl₂O₂ Molecular Weight: 190.82 g/mol |
MDL:
MFCD00013930 CAS-Nummer: 68716-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...