2,4-Dichlorpyridin

Lieferant: COMBI-BLOCKS

OS-7440-5G OS-7440-25G OS-7440-100G
COBBOS-7440-5GEA 25.5 EUR
COBBOS-7440-5G COBBOS-7440-25G COBBOS-7440-100G
2,4-Dichlorpyridin
2,4-Dichloropyridine
Formel: C₅H₃Cl₂N
Molecular Weight: 147,99 g/mol
MDL: MFCD00955618
CAS-Nummer: 26452-80-2
EINECS: 247-717-7
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR