2,4-Dichlorthiobenzamid

Lieferant: FLUOROCHEM

Synonyme: 2,4-Dichlorthiobenzoesäureamid

021316-1G 021316-2.5G
FLUO021316-1GEA 91.2 EUR
FLUO021316-1G FLUO021316-2.5G
2,4-Dichlorthiobenzamid
2,4-Dichlorthiobenzamid
Formel: C₇H₅Cl₂NS
MDL: MFCD00173930
CAS-Nummer: 2775-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR