2,4-Pyridindicarbonitril

Lieferant: CHEMPUR

Synonyme: 2,4-Dicyanpyridin

XBX00038.250 XBX00038.1
CHMPXBX00038.250EA 0 EUR
CHMPXBX00038.250 MAYBXBX00038.1
2,4-Pyridindicarbonitril
2,4-Pyridindicarbonitril
Formel: C₇H₃N₃
Molecular Weight: 129,12 g/mol
Siedepunkt: 274 °C (1013 hPa)
Schmelzpunkt: 91…92 °C
MDL: MFCD00014627
CAS-Nummer: 29181-50-8
EINECS: 249-492-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR