2,4-Difluor-5-nitrobenzoesäure
Lieferant: COMBI-BLOCKS
CA-4001-5G
CA-4001-25G
CA-4001-1G
COBBCA-4001-5GEA
63.9
EUR
COBBCA-4001-5G
COBBCA-4001-25G
COBBCA-4001-1G
2,4-Difluor-5-nitrobenzoesäure
2,4-Difluoro-5-nitrobenzoic acid
Formel:
C₇H₃F₂NO₄ Molecular Weight: 203.1 g/mol |
MDL:
MFCD04115713 CAS-Nummer: 153775-33-8 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...