2,4-Difluorbenzolsulfonamid

Lieferant: CHEMPUR

Synonyme: 2,4-Difluorbenzolsulfonsäureamid

CD10447.250 CD10447.1
CHMPCD10447.250EA 0 EUR
CHMPCD10447.250 MAYBCD10447.1
2,4-Difluorbenzolsulfonamid
2,4-Difluorbenzolsulfonamid
Formel: C₆H₅F₂NO₂S
Molecular Weight: 193,17 g/mol
MDL: MFCD01320743
CAS-Nummer: 13656-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR