2,4-Difluorbenzolsulfonamid

Lieferant: FLUOROCHEM

Synonyme: 2,4-Difluorbenzolsulfonsäureamid

CFF1017.1 005526-1G 005526-5G
CHMPCFF1017.1EA 522 EUR
CHMPCFF1017.1 FLUO005526-1G FLUO005526-5G
2,4-Difluorbenzolsulfonamid
2,4-Difluorbenzolsulfonamid
Formel: C₆H₅F₂NO₂S
Molecular Weight: 193,17 g/mol
MDL: MFCD01320743
CAS-Nummer: 13656-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR