2,4-Difluorphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

BB-2274-5G BB-2274-25G BB-2274-100G
COBBBB-2274-5GEA 31.2 EUR
COBBBB-2274-5G COBBBB-2274-25G COBBBB-2274-100G
2,4-Difluorphenylboronsäure
2,4-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
Molecular Weight: 157,91 g/mol
MDL: MFCD01318998
CAS-Nummer: 144025-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR