2,4-Dihydroxypyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
4-Hydroxy-2-pyridon, Pyridin-2,4-diol
OS-7554-25G
OS-7554-5G
OS-7554-1G
COBBOS-7554-25GEA
88.4
EUR
COBBOS-7554-25G
COBBOS-7554-5G
COBBOS-7554-1G
2,4-Dihydroxypyridin
2,4-Dihydroxypyridin
Formel:
C₅H₅NO₂ Molecular Weight: 111,10 g/mol Siedepunkt: 315 °C (1013 hPa) Schmelzpunkt: 275…277 °C |
MDL:
MFCD00006273 CAS-Nummer: 626-03-9 EINECS: 210-924-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...