2,5-Dibrom-3-picolin

Lieferant: COMBI-BLOCKS

Synonyme: 2,5-Dibrom-3-methylpyridin

OS-7593-100G OS-7593-5G OS-7593-25G
COBBOS-7593-100GEA 181 EUR
COBBOS-7593-100G COBBOS-7593-5G COBBOS-7593-25G
2,5-Dibrom-3-picolin
2,5-Dibrom-3-picolin
Formel: C₆H₅Br₂N
Molecular Weight: 250.92 g/mol
MDL: MFCD02093085
CAS-Nummer: 3430-18-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR