2,5-Dibrom-4-picolin

Lieferant: COMBI-BLOCKS

Synonyme: 2,5-Dibrom-4-methylpyridin

PY-7115-1G PY-7115-5G PY-7115-25G PY-7115-100G
COBBPY-7115-1GEA 22 EUR
COBBPY-7115-1G COBBPY-7115-5G COBBPY-7115-25G COBBPY-7115-100G
2,5-Dibrom-4-picolin
2,5-Dibrom-4-picolin
Formel: C₆H₅Br₂N
Molecular Weight: 250,92 g/mol
MDL: MFCD00234955
CAS-Nummer: 3430-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR