3,6-Dibrom-2-picolin
Lieferant: COMBI-BLOCKS
Synonyme:
3,6-Dibrom-2-methylpyridin
PY-7092-1G
PY-7092-25G
PY-7092-5G
COBBPY-7092-1GEA
0
EUR
COBBPY-7092-1G
COBBPY-7092-25G
COBBPY-7092-5G
3,6-Dibrom-2-picolin
3,6-Dibrom-2-picolin
Formel:
C₆H₅Br₂N Molecular Weight: 250,92 g/mol |
MDL:
MFCD06254589 CAS-Nummer: 39919-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...