2,5-Difluorbenzolsulfonamid

Lieferant: FLUOROCHEM

Synonyme: 2,5-Difluorbenzolsulfonsäureamid

CFF1018.1 005521-1G 005521-5G
CHMPCFF1018.1EA 522 EUR
CHMPCFF1018.1 FLUO005521-1G FLUO005521-5G
2,5-Difluorbenzolsulfonamid
2,5-Difluorbenzolsulfonamid
Formel: C₆H₅F₂NO₂S
Molecular Weight: 193,17 g/mol
MDL: MFCD00798115
CAS-Nummer: 120022-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR