2,5-Difluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2,5-Difluorbenzolboronsäure, (2,5-Difluorphenyl)metaborsäure, Dihydroxy(2,5-difluorphenyl)boran

BB-2663-5G BB-2663-25G BB-2663-100G
COBBBB-2663-5GEA 38.5 EUR
COBBBB-2663-5G COBBBB-2663-25G COBBBB-2663-100G
2,5-Difluorphenylboronsäure
2,5-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
Molecular Weight: 157.91 g/mol
MDL: MFCD01863171
CAS-Nummer: 193353-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR