2,5-Diiod-4-methylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 2,5-Diiod-4-methyl-1H-imidazol

HI-1034-5G HI-1034-1G
COBBHI-1034-5GEA 402 EUR
COBBHI-1034-5G COBBHI-1034-1G
2,5-Diiod-4-methylimidazol
2,5-Diiodo-4-methylimidazole
Formel: C₄H₄I₂N₂
Molecular Weight: 333,9 g/mol
MDL: MFCD02179520
CAS-Nummer: 149510-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR