2,5-Dimethoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2,5-Dimethoxyphenyl)metaborsäure, 2,5-Dimethoxybenzolboronsäure, Dihydroxy(2,5-dimethoxyphenyl)boran

BB-2668-1G BB-2668-5G BB-2668-100G BB-2668-25G
COBBBB-2668-1GEA 25.4 EUR
COBBBB-2668-1G COBBBB-2668-5G COBBBB-2668-100G COBBBB-2668-25G
2,5-Dimethoxyphenylboronsäure
2,5-Dimethoxyphenylboronsäure
Formel: C₈H₁₁BO₄
Molecular Weight: 181.98 g/mol
MDL: MFCD01318181
CAS-Nummer: 107099-99-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR