2,6-Di-tert-butyl-4-methylpyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2,6-Di-Tert-butyl-4-methylpyridine
OS-7542-25G
OS-7542-5G
OS-7542-1G
COBBOS-7542-25GEA
399
EUR
COBBOS-7542-25G
COBBOS-7542-5G
COBBOS-7542-1G
2,6-Di-tert-butyl-4-methylpyridin
2,6-Di-tert-butyl-4-methylpyridin
Formel:
C₁₄H₂₃N Molecular Weight: 205.34 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00006305 CAS-Nummer: 38222-83-2 EINECS: 253-834-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...