2,6-Dichlorphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: (2,6-Dichlorphenyl)metaborsäure, 2,6-Dichlorbenzolboronsäure, Dihydroxy(2,6-dichlorphenyl)boran

BB-2268-5G BB-2268-25G BB-2268-100G
COBBBB-2268-5GEA 25.5 EUR
COBBBB-2268-5G COBBBB-2268-25G COBBBB-2268-100G
2,6-Dichlorphenylboronsäure
2,6-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190.82 g/mol
MDL: MFCD00064869
CAS-Nummer: 73852-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR