2,6-Difluor-4-hydroxybenzaldehyd 95%
Lieferant: Thermo Scientific
|
Achtung
|
H31974.MD
H31974.03
H31974.MDEA
53.5
EUR
H31974.MD
H31974.03
2,6-Difluor-4-hydroxybenzaldehyd 95%
2,6-Difluoro-4-hydroxybenzaldehyde
Purity: 95%
CAS No.: 532967-21-8
Molecular Formula: C7H4F2O2
Formula Weight: 158.11
Melting Point: 179-182°
MDL No.: MFCD07772042
Formel:
C₇H₄F₂O₂ Schmelzpunkt: 179…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07772042 CAS-Nummer: 532967-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...