2,6-Difluor-4-methoxybenzaldehyd

Lieferant: CHEMPUR

10-0015.1 10-0015.5
CHMP10-0015.1EA 0 EUR
CHMP10-0015.1 CHMP10-0015.5
2,6-Difluor-4-methoxybenzaldehyd
2,6-Difluoro-4-methoxybenzaldehyde
Formel: C₈H₆F₂O₂
Molecular Weight: 172.13 g/mol
MDL: MFCD04114368
CAS-Nummer: 256417-10-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR