2,6-Difluorphenylboronsäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
Dihydroxy(2,6-difluorophenyl)borane, 2,6-Difluorobenzeneboronic acid, (2,6-Difluorophenyl)metaboric acid
BB-2659-100G
BB-2659-5G
BB-2659-1G
BB-2659-25G
COBBBB-2659-100GEA
385
EUR
COBBBB-2659-100G
COBBBB-2659-5G
COBBBB-2659-1G
COBBBB-2659-25G
2,6-Difluorphenylboronsäure
2,6-Difluorphenylboronsäure
Formel:
C₆H₅BF₂O₂ Molecular Weight: 157.91 g/mol |
MDL:
MFCD00792436 CAS-Nummer: 162101-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...