2,7-Dibrom-9-fluorenon

Lieferant: COMBI-BLOCKS

Synonyme: 2,7-Dibromo-9H-fluoren-9-one

OT-0555-5G OT-0555-25G OT-0555-100G
COBBOT-0555-5GEA 35.9 EUR
COBBOT-0555-5G COBBOT-0555-25G COBBOT-0555-100G
2,7-Dibrom-9-fluorenon
2,7-Dibrom-9-fluorenon
Formel: C₁₃H₆Br₂O
Molecular Weight: 338 g/mol
MDL: MFCD00010790
CAS-Nummer: 14348-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR