2,7-Dibromfluoren

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 2,7-Dibromo-9H-fluorene

OT-0516-5G OT-0516-25G OT-0516-100G
COBBOT-0516-5GEA 26 EUR
COBBOT-0516-5G COBBOT-0516-25G COBBOT-0516-100G
2,7-Dibromfluoren
2,7-Dibromfluoren
Formel: C₁₃H₈Br₂
Molecular Weight: 324,01 g/mol
MDL: MFCD00019048
CAS-Nummer: 16433-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR