2-Amino-3,5-dichlorpyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
3,5-Dichlorpyridin-2-amin, 3,5-Dichlor-2-pyridylamin
PY-7819-100G
PY-7819-25G
PY-7819-5G
COBBPY-7819-100GEA
226
EUR
COBBPY-7819-100G
COBBPY-7819-25G
COBBPY-7819-5G
2-Amino-3,5-dichlorpyridin
2-Amino-3,5-dichlorpyridin
Formel:
C₅H₄Cl₂N₂ Molecular Weight: 163.01 g/mol Schmelzpunkt: 79…83 °C |
MDL:
MFCD00006313 CAS-Nummer: 4214-74-8 EINECS: 224-143-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...