2-Amino-3,5-dinitro-6-picolin

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dinitro-6-methyl-2-pyridylamin, 2-Amino-3,5-dinitro-6-methylpyridin

PY-7104-1G PY-7104-5G
COBBPY-7104-1GEA 290 EUR
COBBPY-7104-1G COBBPY-7104-5G
2-Amino-3,5-dinitro-6-picolin
2-Amino-3,5-dinitro-6-picolin
Formel: C₆H₆N₄O₄
Molecular Weight: 198,14 g/mol
MDL: MFCD00091867
CAS-Nummer: 25864-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR