2-Amino-3-brom-5-chlor-4-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-5-chlor-4-methyl-2-pyridylamin

PY-1201-1G PY-1201-5G
COBBPY-1201-1GEA 303 EUR
COBBPY-1201-1G COBBPY-1201-5G
2-Amino-3-brom-5-chlor-4-methylpyridin
2-Amino-3-bromo-5-chloro-4-methylpyridine
Formel: C₆H₆BrClN₂
Molecular Weight: 221.48 g/mol
MDL: MFCD13195773
CAS-Nummer: 1199773-28-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR