2-Amino-3-brom-5-methylbenzoesäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A15443.06
A15443.14
A15443.06EA
67.7
EUR
A15443.06
A15443.14
2-Amino-3-brom-5-methylbenzoesäure ≥98%
2-Amino-3-bromo-5-methylbenzoic acid
Purity: 98+%
CAS No.: 13091-43-5
Molecular Formula: C8H8BrNO2
Formula Weight: 230.07
Melting Point: 202-206°
Beilstein Reference: 3540992
Formel:
C₈H₈BrNO₂ Molecular Weight: 230,06 g/mol Schmelzpunkt: 202…206 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051705 CAS-Nummer: 13091-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...