2-Amino-3-brom-5-methylbenzoesäure
Lieferant: CHEMPUR
BTB04576.250
BTB04576.1
CHMPBTB04576.250EA
0
EUR
CHMPBTB04576.250
MAYBBTB04576.1
2-Amino-3-brom-5-methylbenzoesäure
2-Amino-3-bromo-5-methylbenzoic acid
Formel:
C₈H₈BrNO₂ Molecular Weight: 230,06 g/mol |
MDL:
MFCD00051705 CAS-Nummer: 13091-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...