2-Amino-3-brom-5-nitropyridin
Lieferant: COMBI-BLOCKS
Synonyme:
3-Brom-5-nitro-2-pyridylamin, 3-Brom-5-nitropyridin-2-amin
OT-0064-100G
OT-0064-5G
OT-0064-1G
OT-0064-25G
COBBOT-0064-100GEA
260
EUR
COBBOT-0064-100G
COBBOT-0064-5G
COBBOT-0064-1G
COBBOT-0064-25G
2-Amino-3-brom-5-nitropyridin
2-Amino-3-brom-5-nitropyridin
Formel:
C₅H₄BrN₃O₂ Molecular Weight: 218,01 g/mol |
MDL:
MFCD00955628 CAS-Nummer: 15862-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...