2-Amino-3-brompyridin

Lieferant: COMBI-BLOCKS
Achtung

OS-7617-5G OS-7617-25G OS-7617-100G
COBBOS-7617-5GEA 38.5 EUR
COBBOS-7617-5G COBBOS-7617-25G COBBOS-7617-100G
2-Amino-3-brompyridin
2-Amino-3-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
MDL: MFCD03095187
CAS-Nummer: 13534-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR