2-Amino-5-nitro-3-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Methyl-5-nitro-2-pyridylamin, 2-Amino-3-methyl-5-nitropyridin

M-Clarity™ Qualitätsstufe = MQ100

703974-1G 703974-5G
SIAL703974-1GEA 34.7 EUR
SIAL703974-1G SIAL703974-5G
2-Amino-5-nitro-3-picolin, Sigma-Aldrich®
2-Amino-5-nitro-3-picolin
Formel: C₆H₇N₃O₂
Molecular Weight: 153,14 g/mol
MDL: MFCD03095060
CAS-Nummer: 18344-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR