2-Amino-5-nitro-4-picolin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Methyl-5-nitro-2-pyridylamin, 2-Amino-4-methyl-5-nitropyridin

L19729.06 L19729.14
L19729.06EA 90.1 EUR
L19729.06 L19729.14
2-Amino-5-nitro-4-picolin 98%
2-Amino-5-nitro-4-picolin

Purity: 98% CAS No.: 21901-40-6 Molecular Formula: C6H7N3O2 Formula Weight: 153.14 Melting Point: 223-225° Beilstein Reference: 137695

Formel: C₆H₇N₃O₂
Molecular Weight: 153,14 g/mol
Schmelzpunkt: 223…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010692
CAS-Nummer: 21901-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR