2-Amino-5-brom-3-methylpyridin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 2-Amino-5-brom-3-picolin, 5-Brom-3-methyl-2-pyridylamin, 5-Brom-3-methylpyridin-2-amin

PY-1117-5G PY-1117-25G PY-1117-100G
COBBPY-1117-5GEA 26.2 EUR
COBBPY-1117-5G COBBPY-1117-25G COBBPY-1117-100G
2-Amino-5-brom-3-methylpyridin
2-Amino-5-brom-3-methylpyridin
Formel: C₆H₇BrN₂
Molecular Weight: 187,04 g/mol
MDL: MFCD00068232
CAS-Nummer: 3430-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR