2-Amino-5-bromnicotinsäure

Lieferant: COMBI-BLOCKS

CA-4157-5G CA-4157-25G CA-4157-1G
COBBCA-4157-5GEA 50.8 EUR
COBBCA-4157-5G COBBCA-4157-25G COBBCA-4157-1G
2-Amino-5-bromnicotinsäure
2-Amino-5-bromnicotinsäure
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217.02 g/mol
MDL: MFCD01860227
CAS-Nummer: 52833-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR