2-Amino-5-brom-4-methylpyridin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 5-Bromo-4-methylpyridin-2-amine, 5-Bromo-4-methyl-2-pyridylamine, 2-Amino-5-bromo-4-picoline

OT-0052-5G OT-0052-25G OT-0052-1G OT-0052-100G
COBBOT-0052-5GEA 26.2 EUR
COBBOT-0052-5G COBBOT-0052-25G COBBOT-0052-1G COBBOT-0052-100G
2-Amino-5-brom-4-methylpyridin
2-Amino-5-brom-4-methylpyridin
Formel: C₆H₇BrN₂
Molecular Weight: 187,04 g/mol
MDL: MFCD03427660
CAS-Nummer: 98198-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR