2-Amino-5-brombenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-5-brombenzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ100

682470-1G 682470-250MG
SIAL682470-1GEA 74.4 EUR
SIAL682470-1G SIAL682470-250MG
2-Amino-5-brombenzolsulfonamid, Sigma-Aldrich®
2-Amino-5-brombenzolsulfonamid
Formel: C₆H₇BrN₂O₂S
Molecular Weight: 251,1 g/mol
MDL: MFCD03426835
CAS-Nummer: 54734-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR