2-Amino-5-chlor-3-fluorpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Chlor-3-fluor-2-pyridylamin, 5-Chlor-3-fluorpyridin-2-amin

PY-7242-1G PY-7242-25G PY-7242-5G
COBBPY-7242-1GEA 26 EUR
COBBPY-7242-1G COBBPY-7242-25G COBBPY-7242-5G
2-Amino-5-chlor-3-fluorpyridin
2-Amino-5-chloro-3-fluoropyridine
Formel: C₅H₄ClFN₂
Molecular Weight: 146,55 g/mol
MDL: MFCD09878434
CAS-Nummer: 246847-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR