2-Amino-5-fluorpyridin

Lieferant: COMBI-BLOCKS

PY-7132-25G PY-7132-5G PY-7132-100G
COBBPY-7132-25GEA 292 EUR
COBBPY-7132-25G COBBPY-7132-5G COBBPY-7132-100G
2-Amino-5-fluorpyridin
2-Amino-5-fluorpyridin
Formel: C₅H₅FN₂
Molecular Weight: 112,11 g/mol
MDL: MFCD01861120
CAS-Nummer: 21717-96-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR