2-Amino-6-brompyridin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 6-Brompyridin-2-amin, 6-Brom-2-pyridylamin

OS-7401-5G OS-7401-25G OS-7401-100G
COBBOS-7401-5GEA 28.5 EUR
COBBOS-7401-5G COBBOS-7401-25G COBBOS-7401-100G
2-Amino-6-brompyridin
2-Amino-6-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
MDL: MFCD00137843
CAS-Nummer: 19798-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR