2-Amino-6-chlorpyridin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 6-Chlorpyridin-2-amin, 6-Chlor-2-pyridylamin

OS-7520-5G OS-7520-25G
COBBOS-7520-5GEA 26.2 EUR
COBBOS-7520-5G COBBOS-7520-25G
2-Amino-6-chlorpyridin
2-Amino-6-chlorpyridin
Formel: C₅H₅ClN₂
Molecular Weight: 128,56 g/mol
MDL: MFCD00234068
CAS-Nummer: 45644-21-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR