2-Amino-6-ethylpyridin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 6-Ethylpyridin-2-amin, 6-Ethyl-2-pyridylamin

A19146.09 A19146.88
A19146.09EA 166 EUR
A19146.09 A19146.88
2-Amino-6-ethylpyridin 97%
2-Amino-6-ethylpyridine

Purity: 97% CAS No.: 21717-29-3 Molecular Formula: C7H10N2 Formula Weight: 122.17 Refractive Index: 1.5680 Odour: Unpleasant UN No.: 2810 MDL No.: MFCD00218265

Formel: C₇H₁₀N₂
Molecular Weight: 122.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00218265
CAS-Nummer: 21717-29-3
EINECS: 244-540-7
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR