2-Amino-6-fluorbenzothiazol
Lieferant: COMBI-BLOCKS
Synonyme:
6-Fluorbenzothiazol-2-ylamin, 6-Fluor-1,3-benzothiazol-2-amin
HC-2256-5G
HC-2256-25G
HC-2256-100G
COBBHC-2256-5GEA
26.2
EUR
COBBHC-2256-5G
COBBHC-2256-25G
COBBHC-2256-100G
2-Amino-6-fluorbenzothiazol
2-Amino-6-fluorobenzothiazole
Formel:
C₇H₅FN₂S Molecular Weight: 168.19 g/mol Siedepunkt: 304 Schmelzpunkt: 183 Dichte: 1.59 |
MDL:
MFCD00013336 CAS-Nummer: 348-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...